Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177759.3 | NP_808427.1 | 4 - 5 | 901 | 73 | |
GenBank mRNA | AK082047.1 | - | 4 - 5 | 902 | 73 | |
BC075662.1 | - | 4 - 5 | 880 | 73 | ||
BC150892.1 | - | 4 - 5 | 691 | 9087803 | 73 | |
BC172040.1 | - | 4 - 5 | 689 | 9054326 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |