Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177759.3 | NP_808427.1 | 10 - 11 | 1537 | 70 | |
GenBank mRNA | AK082047.1 | - | 10 - 11 | 1538 | 70 | |
BC060151.1 | - | 52 | 70 | |||
BC150892.1 | - | 10 - 11 | 1327 | 9087803 | 70 | |
BC172040.1 | - | 10 - 11 | 1325 | 9054326 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |