Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177781.4 | NP_808449.1 | 11 - 12 | 1388 | 126 | |
GenBank mRNA | AK045771.1 | - | 11 - 12 | 1389 | 126 | |
AY231177.1 | - | 11 - 12 | 1361 | 126 | ||
BC120563.1 | - | 11 - 12 | 1379 | 40129486 | 126 | |
BC131963.1 | - | 11 - 12 | 1379 | 40130240 | 126 | |
KF460461.1 | - | 11 - 12 | 1361 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |