Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177839.3 | NP_808507.2 | 7 - 8 | 1716 | 66 | |
GenBank mRNA | AB052768.1 | - | 2 - 3 | 242 | 66 | |
AF455756.1 | - | 7 - 8 | 1683 | 66 | ||
AJ580920.1 | - | 6 - 7 | 1635 | 66 | ||
BC138335.1 | - | 7 - 8 | 1677 | 8861355 | 66 | |
BC138336.1 | - | 7 - 8 | 1677 | 8861356 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |