Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hapln4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177900.4 | NP_808568.1 | 2 - 3 | 155 | 118 | |
GenBank mRNA | AB107882.1 | - | 2 - 3 | 155 | 118 | |
AK034300.1 | - | 2 - 3 | 164 | 118 | ||
AK158408.1 | - | 2 - 3 | 182 | 118 | ||
AY262758.1 | - | 2 - 3 | 164 | 118 | ||
AY269789.1 | - | 2 - 3 | 227 | 118 | ||
BC125339.1 | - | 2 - 3 | 142 | 40129854 | 118 | |
BC125341.1 | - | 2 - 3 | 142 | 40129856 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |