Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033500.3 | NP_001028672.2 | 1 - 2 | 364 | 74 | |
GenBank mRNA | AK052055.1 | - | 1 - 2 | 368 | 74 | |
AK135824.1 | - | 1 - 2 | 344 | 74 | ||
AK142559.1 | - | 1 - 2 | 344 | 74 | ||
BB658361.1 | - | 1 - 2 | 364 | 74 | ||
BC145956.1 | - | 1 - 2 | 250 | 74 | ||
BC145958.1 | - | 1 - 2 | 249 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |