Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178253.5 | NP_839984.1 | 7 - 8 | 738 | 138 | |
GenBank mRNA | AK144621.1 | - | 7 - 8 | 678 | 138 | |
AY238938.1 | - | 7 - 8 | 738 | 138 | ||
BC119567.2 | - | 7 - 8 | 716 | 40057793 | 138 | |
BC119568.2 | - | 7 - 8 | 722 | 40057794 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |