Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178743.3 | NP_848858.2 | 7 - 8 | 985 | 81 | |
GenBank mRNA | AF345196.1 | - | 7 - 8 | 985 | 81 | |
AK052202.1 | - | 2 - 3 | 1072 | 81 | ||
AK078545.1 | - | 6 - 7 | 2065 | 81 | ||
AK089182.1 | - | 7 - 8 | 965 | 81 | ||
BC038604.1 | - | 7 - 8 | 978 | 81 | ||
BC132493.1 | - | 2 - 3 | 853 | 40130770 | 81 | |
BC137897.1 | - | 2 - 3 | 853 | 8860916 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |