Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178758.3 | NP_848873.1 | 9 - 10 | 1390 | 127 | |
GenBank mRNA | AK050132.1 | - | 10 - 11 | 1487 | 127 | |
AK050219.1 | - | 9 - 10 | 1415 | 127 | ||
AK050288.1 | - | 9 - 10 | 1410 | 127 | ||
AK050404.1 | - | 9 - 10 | 1450 | 127 | ||
AK050458.1 | - | 9 - 10 | 1352 | 127 | ||
AK149591.1 | - | 9 - 10 | 1410 | 127 | ||
BC095985.1 | - | 9 - 10 | 1390 | 4971395 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |