Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252458.1 | NP_001239387.1 | 12 - 13 | 1904 | 76 | |
NM_178900.4 | NP_849231.1 | 11 - 12 | 2189 | 76 | ||
GenBank mRNA | AK037030.1 | - | 11 - 12 | 2190 | 76 | |
AK154029.1 | - | 11 - 12 | 2038 | 76 | ||
AK170236.1 | - | 12 - 13 | 1877 | 76 | ||
AK170761.1 | - | 12 - 13 | 1868 | 76 | ||
BC095949.1 | - | 12 - 13 | 1854 | 4950957 | 76 | |
BC096444.1 | - | 12 - 13 | 1872 | 4216827 | 76 | |
BF146730.1 | - | 3 - 4 | 330 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |