Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Canx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110499.1 | NP_001103969.1 | 10 - 11 | 1366 | 121 | |
NM_001110500.1 | NP_001103970.1 | 10 - 11 | 1334 | 121 | ||
NM_007597.3 | NP_031623.1 | 10 - 11 | 1370 | 121 | ||
GenBank mRNA | AK028279.1 | - | 10 - 11 | 1298 | 121 | |
AK084175.1 | - | 10 - 11 | 1310 | 121 | ||
AK159298.1 | - | 10 - 11 | 1354 | 121 | ||
AK165508.1 | - | 10 - 11 | 1269 | 121 | ||
AK166649.1 | - | 10 - 11 | 1347 | 121 | ||
BC012408.1 | - | 10 - 11 | 1288 | 3497769 | 121 | |
BC040244.1 | - | 10 - 11 | 1297 | 2582119 | 121 | |
L18888.1 | - | 9 - 10 | 1182 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |