Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Canx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110499.1 | NP_001103969.1 | 6 - 7 | 714 | 130 | |
NM_001110500.1 | NP_001103970.1 | 6 - 7 | 682 | 130 | ||
NM_007597.3 | NP_031623.1 | 6 - 7 | 718 | 130 | ||
GenBank mRNA | AK028279.1 | - | 6 - 7 | 646 | 130 | |
AK084175.1 | - | 6 - 7 | 658 | 130 | ||
AK159298.1 | - | 6 - 7 | 702 | 130 | ||
AK165508.1 | - | 6 - 7 | 617 | 130 | ||
AK166649.1 | - | 6 - 7 | 695 | 130 | ||
BC012408.1 | - | 6 - 7 | 636 | 3497769 | 130 | |
BC040244.1 | - | 6 - 7 | 645 | 2582119 | 130 | |
L18888.1 | - | 5 - 6 | 530 | 130 | ||
L23865.1 | - | 5 - 6 | 470 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |