Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019810.4 | NP_062784.3 | 10 - 11 | 1370 | 61 | |
GenBank mRNA | AF208031.1 | - | 10 - 11 | 1370 | 61 | |
AK033512.1 | - | 10 - 11 | 1356 | 61 | ||
AK085602.1 | - | 10 - 11 | 1161 | 61 | ||
BC003845.1 | - | 10 - 11 | 1350 | 3497611 | 61 | |
U27269.1 | - | 8 - 9 | 940 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |