Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sectm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026907.3 | NP_081183.3 | 2 - 3 | 166 | 92 | |
GenBank mRNA | AK029082.1 | - | 2 - 3 | 167 | 92 | |
AK143807.1 | - | 2 - 3 | 150 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |