Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133847.3 | NP_598608.2 | 6 - 7 | 909 | 60 | |
GenBank mRNA | AB093220.2 | - | 5 - 6 | 653 | 60 | |
AK035622.1 | - | 6 - 7 | 871 | 60 | ||
AK048158.1 | - | 6 - 7 | 667 | 60 | ||
AK083541.1 | - | 6 - 7 | 909 | 60 | ||
AK087048.1 | - | 6 - 7 | 662 | 60 | ||
AK088012.1 | - | 6 - 7 | 665 | 60 | ||
AK162435.1 | - | 6 - 7 | 670 | 60 | ||
AK171487.1 | - | 6 - 7 | 667 | 60 | ||
BC110309.1 | - | 6 - 7 | 665 | 5324218 | 60 | |
BC116320.2 | - | 6 - 7 | 861 | 40096002 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |