Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039200.2 | NP_001034289.1 | 7 - 8 | 824 | 84 | |
NM_146157.4 | NP_666269.2 | 7 - 8 | 824 | 84 | ||
GenBank mRNA | AK049061.1 | - | 7 - 8 | 815 | 84 | |
AK077496.1 | - | 7 - 8 | 814 | 84 | ||
AK089289.1 | - | 7 - 8 | 803 | 84 | ||
AK129052.1 | - | 7 - 8 | 777 | 84 | ||
AK138637.1 | - | 7 - 8 | 796 | 84 | ||
AK161677.1 | - | 7 - 8 | 787 | 84 | ||
AK170739.1 | - | 7 - 8 | 803 | 84 | ||
BC048948.1 | - | 10 - 11 | 84 | |||
BC056952.1 | - | 2 - 3 | 272 | 84 | ||
BC129804.1 | - | 7 - 8 | 811 | 40087024 | 84 | |
BC139366.1 | - | 7 - 8 | 810 | 8862388 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |