Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnkd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039509.1 | NP_001034598.1 | 1 - 2 | 103 | 102 | |
NM_025580.2 | NP_079856.2 | 1 - 2 | 103 | 102 | ||
GenBank mRNA | AF318058.1 | - | 1 - 2 | 207 | 102 | |
AK009545.1 | - | 1 - 2 | 80 | 102 | ||
AK012976.1 | - | 1 - 2 | 82 | 102 | ||
AK173125.1 | - | 1 - 2 | 77 | 102 | ||
AY039041.1 | - | 1 - 2 | 66 | 102 | ||
AY299972.1 | - | 1 - 2 | 66 | 102 | ||
BC008274.1 | - | 1 - 2 | 112 | 102 | ||
BC058945.1 | - | 1 - 2 | 82 | 6812127 | 102 | |
BC116236.1 | - | 1 - 2 | 190 | 102 | ||
BC116237.1 | - | 1 - 2 | 190 | 102 | ||
CK623847.1 | - | 1 - 2 | 73 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |