Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290783.1 | NP_001277712.1 | 5 - 6 | 1324 | 81 | |
NM_026137.5 | NP_080413.2 | 5 - 6 | 1358 | 81 | ||
NM_028542.1 | NP_082818.1 | 5 - 6 | 1610 | 81 | ||
GenBank mRNA | AF220146.1 | - | 4 - 5 | 880 | 81 | |
AF353243.1 | - | 5 - 6 | 1004 | 81 | ||
AF513713.1 | - | 5 - 6 | 827 | 81 | ||
AK006837.1 | - | 4 - 5 | 846 | 81 | ||
AK017549.1 | - | 5 - 6 | 940 | 81 | ||
BC057366.1 | - | 5 - 6 | 1324 | 81 | ||
BC080853.1 | - | 5 - 6 | 1273 | 81 | ||
BC108332.1 | - | 5 - 6 | 1114 | 6493226 | 81 | |
BC120553.1 | - | 5 - 6 | 861 | 40129476 | 81 | |
BC125287.1 | - | 5 - 6 | 860 | 40129802 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |