Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10570695 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570694 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570708 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570661 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570733 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570662 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570653 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570734 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570673 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570719 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570713 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570696 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570718 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10570693 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10570732 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10570660 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10570717 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10570693 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10570660 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10570717 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10570732 |
- |
- |
Mouse Genome 430 2.0 Array |
1450709_at |
- |
- |
Mouse Genome 430 2.0 Array |
1450709_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0800001918.mm.1 |
NM_001177528.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001918.mm.1 |
NM_001177528.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001918.mm.1 |
TC0800000252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001844.mm.1 |
NM_007851.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001844.mm.1 |
NM_007851.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001844.mm.1 |
TC0800000229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001848.mm.1 |
NM_007851.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001848.mm.1 |
NM_007851.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001848.mm.1 |
TC0800000229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001919.mm.1 |
NM_001177528.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001919.mm.1 |
NM_001177528.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001919.mm.1 |
TC0800000252.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001846.mm.1 |
NM_007851.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001846.mm.1 |
NM_007851.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001846.mm.1 |
TC0800000229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001847.mm.1 |
NM_007851.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001847.mm.1 |
NM_007851.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001847.mm.1 |
TC0800000229.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1450709_at |
- |
- |
Mouse Expression Set 430 |
1450709_PM_at |
- |
- |
Mouse Expression Set 430 |
1450709_at |
- |
- |
Mouse Expression Set 430 |
1450709_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1450709_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1450709_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1450709_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1450709_at |
- |
- |