Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK144562.1 | - | 2 | 1100 | 97 | |
X90829.1 | - | 1 | 2385 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon |