Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027649.3 | NP_081925.2 | 6 - 7 | 794 | 90 | |
GenBank mRNA | AK005865.1 | - | 6 - 7 | 795 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |