Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nagpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013796.3 | NP_038824.2 | 1 - 2 | 108 | 76 | |
GenBank mRNA | AK032158.1 | - | 1 - 2 | 109 | 76 | |
AK087468.1 | - | 1 - 2 | 108 | 76 | ||
AK138035.1 | - | 1 - 2 | 101 | 76 | ||
AK155450.1 | - | 1 - 2 | 87 | 76 | ||
AK159629.1 | - | 1 - 2 | 87 | 76 | ||
AK162633.1 | - | 1 - 2 | 90 | 76 | ||
AK170219.1 | - | 1 - 2 | 90 | 76 | ||
BC039790.1 | - | 1 - 2 | 100 | 4951805 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |