Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172073.1 | NP_001165544.1 | 6 - 7 | 761 | 106 | |
NM_028209.2 | NP_082485.1 | 6 - 7 | 760 | 106 | ||
GenBank mRNA | AK005923.1 | - | 6 - 7 | 761 | 106 | |
AK133931.1 | - | 6 - 7 | 725 | 106 | ||
AK184523.1 | - | 3 - 4 | 236 | 106 | ||
BC025435.1 | - | 6 - 7 | 709 | 5004463 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |