Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028636.2 | NP_082912.1 | 24 - 25 | 2896 | 57 | |
GenBank mRNA | AK036765.1 | - | 1932 | 57 | ||
AK049370.1 | - | 24 - 25 | 2917 | 57 | ||
AK153935.1 | - | 24 - 25 | 2917 | 57 | ||
AK163077.1 | - | 24 - 25 | 2898 | 57 | ||
BC016253.1 | - | 24 - 25 | 2896 | 4911979 | 57 | |
BC023038.1 | - | 5 - 6 | 745 | 57 | ||
BC039913.1 | - | 8 - 9 | 1061 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |