Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163579.1 | NP_001157051.1 | 2 - 3 | 445 | 137 | |
NM_001163580.1 | NP_001157052.1 | 2 - 3 | 445 | 137 | ||
NM_028915.3 | NP_083191.2 | 2 - 3 | 445 | 137 | ||
GenBank mRNA | AK004646.1 | - | 2 - 3 | 446 | 137 | |
AK016559.1 | - | 2 - 3 | 384 | 137 | ||
AK029563.1 | - | 2 - 3 | 362 | 137 | ||
AK173256.1 | - | 2 - 3 | 373 | 137 | ||
BC061202.1 | - | 2 - 3 | 401 | 137 | ||
BC099680.1 | - | 2 - 3 | 409 | 137 | ||
BC138734.1 | - | 2 - 3 | 419 | 8861756 | 137 | |
BC138735.1 | - | 2 - 3 | 419 | 8861757 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |