Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168403.1 | NP_001161875.1 | 5 - 6 | 1000 | 127 | |
NM_001168404.1 | NP_001161876.1 | 5 - 6 | 537 | 127 | ||
NM_001168405.1 | NP_001161877.1 | 4 - 5 | 894 | 127 | ||
NM_029898.3 | NP_084174.2 | 4 - 5 | 894 | 127 | ||
GenBank mRNA | AK039553.1 | - | 4 - 5 | 895 | 127 | |
AK045480.1 | - | 4 - 5 | 528 | 127 | ||
AK081197.1 | - | 5 - 6 | 538 | 127 | ||
AK138273.1 | - | 4 - 5 | 549 | 127 | ||
AK140417.1 | - | 5 - 6 | 864 | 127 | ||
AK155092.1 | - | 3 - 4 | 335 | 127 | ||
AK171119.1 | - | 3 - 4 | 335 | 127 | ||
BC152317.1 | - | 4 - 5 | 521 | 40088531 | 127 | |
BC152536.1 | - | 4 - 5 | 521 | 40088530 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |