Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130888.1 | NP_570958.1 | 13 - 14 | 1129 | 143 | |
GenBank mRNA | AB178901.1 | - | 14 - 15 | 1268 | 143 | |
AY260550.1 | - | 13 - 14 | 1129 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |