Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130888.1 | NP_570958.1 | 19 - 20 | 1575 | 96 | |
GenBank mRNA | AB178901.1 | - | 20 - 21 | 1714 | 96 | |
AJ305317.1 | - | 15 - 16 | 1136 | 96 | ||
AJ305318.1 | - | 16 - 17 | 1244 | 96 | ||
AY260550.1 | - | 19 - 20 | 1575 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |