Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166597.1 | NP_001160069.1 | 6 - 7 | 1224 | 56 | |
NM_138306.2 | NP_612179.2 | 5 - 6 | 623 | 56 | ||
GenBank mRNA | AK087503.1 | - | 6 - 7 | 1225 | 56 | |
AK170936.1 | - | 2 - 3 | 246 | 56 | ||
AK210058.1 | - | 1 - 2 | 35 | 56 | ||
BC016217.1 | - | 5 - 6 | 1570 | 56 | ||
BC049228.1 | - | 5 - 6 | 659 | 3668704 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |