Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145588.1 | NP_663563.1 | 6 - 7 | 994 | 73 | |
GenBank mRNA | AK075922.1 | - | 6 - 7 | 985 | 73 | |
AK084310.1 | - | 6 - 7 | 982 | 73 | ||
AK088033.1 | - | 3 - 4 | 570 | 73 | ||
BC003427.1 | - | 6 - 7 | 994 | 2615715 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |