Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028639.3 | NP_082915.1 | 11 - 12 | 1734 | 65 | |
GenBank mRNA | AK040578.1 | - | 11 - 12 | 1737 | 65 | |
AK040610.1 | - | 11 - 12 | 1705 | 65 | ||
AK050263.1 | - | 11 - 12 | 1721 | 65 | ||
AK080284.1 | - | 11 - 12 | 1713 | 65 | ||
AK153704.1 | - | 11 - 12 | 1716 | 65 | ||
AK165244.1 | - | 4 - 5 | 396 | 65 | ||
BC023773.1 | - | 7 - 8 | 692 | 65 | ||
BC042512.1 | - | 11 - 12 | 1411 | 4223801 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |