Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028639.3 | NP_082915.1 | 7 - 8 | 1338 | 68 | |
GenBank mRNA | AK040578.1 | - | 7 - 8 | 1341 | 68 | |
AK040610.1 | - | 7 - 8 | 1309 | 68 | ||
AK050263.1 | - | 7 - 8 | 1325 | 68 | ||
AK080284.1 | - | 7 - 8 | 1317 | 68 | ||
AK153704.1 | - | 7 - 8 | 1320 | 68 | ||
BC023773.1 | - | 3 - 4 | 296 | 68 | ||
BC042512.1 | - | 7 - 8 | 1015 | 4223801 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |