Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172806.2 | NP_766394.2 | 5 - 6 | 1827 | 94 | |
GenBank mRNA | AK122522.1 | - | 3 - 4 | 1477 | 94 | |
BC041669.1 | - | 5 - 6 | 1827 | 3482657 | 94 | |
BC057303.1 | - | 4 - 5 | 1546 | 5702276 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |