Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172806.2 | NP_766394.2 | 6 - 7 | 1902 | 109 | |
GenBank mRNA | AK122522.1 | - | 4 - 5 | 1552 | 109 | |
BC041669.1 | - | 6 - 7 | 1902 | 3482657 | 109 | |
BC057303.1 | - | 5 - 6 | 1621 | 5702276 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |