Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174852.3 | NP_777277.2 | 11 - 12 | 2847 | 62 | |
GenBank mRNA | AK129383.1 | - | 8 - 9 | 2142 | 62 | |
AK155354.1 | - | 11 - 12 | 2888 | 62 | ||
BC043080.1 | - | 11 - 12 | 2450 | 6400617 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |