Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174852.3 | NP_777277.2 | 5 - 6 | 1327 | 73 | |
GenBank mRNA | AK089226.1 | - | 5 - 6 | 1391 | 73 | |
AK129383.1 | - | 2 - 3 | 474 | 73 | ||
AK141796.1 | - | 5 - 6 | 1231 | 73 | ||
AK155354.1 | - | 5 - 6 | 1368 | 73 | ||
AK170155.1 | - | 5 - 6 | 1415 | 73 | ||
AK170187.1 | - | 4 - 5 | 622 | 73 | ||
BC043080.1 | - | 5 - 6 | 930 | 6400617 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |