Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ablim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177696.1 | NP_001171167.1 | 2 - 3 | 331 | 77 | |
NM_001177697.1 | NP_001171168.1 | 2 - 3 | 331 | 77 | ||
NM_001177698.1 | NP_001171169.1 | 2 - 3 | 331 | 77 | ||
NM_001177699.1 | NP_001171170.1 | 2 - 3 | 331 | 77 | ||
NM_001177700.1 | NP_001171171.1 | 2 - 3 | 331 | 77 | ||
NM_177678.7 | NP_808346.3 | 2 - 3 | 331 | 77 | ||
GenBank mRNA | AJ748602.1 | - | 2 - 3 | 287 | 77 | |
AJ748603.1 | - | 2 - 3 | 287 | 77 | ||
AK046243.1 | - | 2 - 3 | 282 | 77 | ||
AK046879.1 | - | 2 - 3 | 280 | 77 | ||
AK082707.1 | - | 2 - 3 | 281 | 77 | ||
AK154494.1 | - | 2 - 3 | 333 | 77 | ||
AY274116.1 | - | 2 - 3 | 258 | 77 | ||
BC141125.1 | - | 2 - 3 | 191 | 9055737 | 77 | |
DQ413175.1 | - | 1 - 2 | 160 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |