Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zranb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017381.2 | NP_059077.1 | 4 - 5 | 319 | 149 | |
GenBank mRNA | AK146275.1 | - | 4 - 5 | 317 | 149 | |
AK146854.1 | - | 4 - 5 | 311 | 149 | ||
AK150291.1 | - | 4 - 5 | 358 | 149 | ||
AK152536.1 | - | 4 - 5 | 358 | 149 | ||
AK160862.1 | - | 3 - 4 | 207 | 149 | ||
AK166534.1 | - | 4 - 5 | 314 | 149 | ||
AK167756.1 | - | 4 - 5 | 317 | 149 | ||
AK167766.1 | - | 4 - 5 | 347 | 149 | ||
AK167862.1 | - | 4 - 5 | 347 | 149 | ||
AK167983.1 | - | 4 - 5 | 347 | 149 | ||
AK168033.1 | - | 4 - 5 | 317 | 149 | ||
AK168162.1 | - | 4 - 5 | 318 | 149 | ||
AK168613.1 | - | 4 - 5 | 349 | 149 | ||
AK169114.1 | - | 4 - 5 | 311 | 149 | ||
AK169335.1 | - | 4 - 5 | 320 | 149 | ||
BC049115.1 | - | 4 - 5 | 329 | 149 | ||
BC132547.1 | - | 4 - 5 | 316 | 40130824 | 149 | |
BC138575.1 | - | 4 - 5 | 316 | 8861595 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |