Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290818.1 | NP_001277747.1 | 8 - 9 | 854 | 57 | |
NM_130863.2 | NP_570933.1 | 9 - 10 | 926 | 57 | ||
GenBank mRNA | AF333028.1 | - | 8 - 9 | 646 | 57 | |
AK155691.1 | - | 8 - 9 | 855 | 57 | ||
AK193363.1 | - | 3 - 4 | 167 | 57 | ||
BC003196.1 | - | 8 - 9 | 642 | 57 | ||
BC033272.1 | - | 7 - 8 | 970 | 57 | ||
BC053922.1 | - | 9 - 10 | 659 | 6309555 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |