Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291818.1 | NP_001278747.1 | 12 - 13 | 1686 | 107 | |
NM_010117.2 | NP_034247.2 | 12 - 13 | 1805 | 107 | ||
GenBank mRNA | AK144437.1 | - | 12 - 13 | 1674 | 107 | |
AK220566.1 | - | 8 - 9 | 3304 | 107 | ||
BC023469.1 | - | 7 - 8 | 953 | 107 | ||
BC027346.1 | - | 12 - 13 | 1750 | 4011421 | 107 | |
M99623.1 | - | 2 - 3 | 126 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |