Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025662.5 | NP_079938.1 | 8 - 9 | 844 | 126 | |
NM_178016.3 | NP_821135.1 | 9 - 10 | 985 | 126 | ||
GenBank mRNA | AK013853.1 | - | 8 - 9 | 841 | 126 | |
AK037552.1 | - | 4 - 5 | 428 | 127 | ||
AK043033.1 | - | 833 | 126 | |||
AK046246.1 | - | 9 - 10 | 973 | 126 | ||
AK077893.1 | - | 8 - 9 | 845 | 126 | ||
AK082806.1 | - | 8 - 9 | 837 | 126 | ||
AK168666.1 | - | 826 | 126 | |||
BC023256.1 | - | 8 - 9 | 826 | 126 | ||
BC047203.1 | - | 4 - 5 | 382 | 126 | ||
BC060175.1 | - | 8 - 9 | 830 | 6827521 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |