Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025949.3 | NP_080225.2 | 14 - 15 | 1452 | 151 | |
GenBank mRNA | AK049349.1 | - | 12 - 13 | 1203 | 151 | |
BC054113.1 | - | 12 - 13 | 1125 | 30015425 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |