Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400034449.mm.1 |
TC0400004119.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400034449.mm.1 |
NM_001177995.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034449.mm.1 |
NM_001177995.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034449.mm.1 |
NM_027504.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034449.mm.1 |
NM_027504.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034443.mm.1 |
TC0400004119.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400034443.mm.1 |
NM_001177995.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034443.mm.1 |
NM_001177995.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034443.mm.1 |
NM_027504.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034443.mm.1 |
NM_027504.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034447.mm.1 |
TC0400004119.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400034447.mm.1 |
NM_001177995.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034447.mm.1 |
NM_001177995.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034447.mm.1 |
NM_027504.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034447.mm.1 |
NM_027504.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034450.mm.1 |
TC0400004119.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400034450.mm.1 |
NM_001177995.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034450.mm.1 |
NM_001177995.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034450.mm.1 |
NM_027504.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034450.mm.1 |
NM_027504.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034451.mm.1 |
TC0400004119.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400034451.mm.1 |
NM_001177995.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034451.mm.1 |
NM_001177995.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034451.mm.1 |
NM_027504.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034451.mm.1 |
NM_027504.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034441.mm.1 |
TC0400004119.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400034441.mm.1 |
NM_001177995.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034441.mm.1 |
NM_001177995.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034441.mm.1 |
NM_027504.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034441.mm.1 |
NM_027504.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034452.mm.1 |
TC0400004119.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400034452.mm.1 |
NM_001177995.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034452.mm.1 |
NM_001177995.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034452.mm.1 |
NM_027504.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400034452.mm.1 |
NM_027504.3 |
Refseq |