Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027982.2 | NP_082258.2 | 2 - 3 | 472 | 83 | |
GenBank mRNA | AK009235.1 | - | 2 - 3 | 411 | 83 | |
AY184802.1 | - | 2 - 3 | 472 | 83 | ||
BC117498.1 | - | 2 - 3 | 468 | 40106734 | 83 | |
BC121788.1 | - | 2 - 3 | 467 | 40106732 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |