Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028385.1 | NP_082661.1 | 17 - 18 | 2916 | 94 | |
GenBank mRNA | AK049143.1 | - | 11 - 12 | 1994 | 94 | |
AK053541.1 | - | 2 - 3 | 1047 | 94 | ||
AK122551.1 | - | 17 - 18 | 2916 | 94 | ||
BC083184.1 | - | 16 - 17 | 2797 | 30536831 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |