Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028385.1 | NP_082661.1 | 20 - 21 | 3815 | 62 | |
GenBank mRNA | AK053541.1 | - | 5 - 6 | 1946 | 62 | |
AK122551.1 | - | 20 - 21 | 3815 | 62 | ||
BC083184.1 | - | 19 - 20 | 3696 | 30536831 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |