Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Antxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054041.2 | NP_473382.1 | 14 - 15 | 1417 | 89 | |
GenBank mRNA | AF378762.1 | - | 14 - 15 | 1357 | 89 | |
AK013005.1 | - | 12 - 13 | 824 | 89 | ||
AK031465.1 | - | 14 - 15 | 1407 | 89 | ||
BC057043.1 | - | 13 - 14 | 881 | 89 | ||
BC094544.1 | - | 14 - 15 | 1362 | 6406827 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |