Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133889.2 | NP_598650.2 | 4 - 5 | 379 | 64 | |
GenBank mRNA | AK037201.1 | - | 4 - 5 | 375 | 64 | |
AK084857.1 | - | 3 - 4 | 1851 | 64 | ||
AK144053.1 | - | 4 - 5 | 374 | 64 | ||
BC021480.1 | - | 3 - 4 | 359 | 64 | ||
BC049111.1 | - | 4 - 5 | 379 | 6415574 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |