Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc41a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173865.3 | NP_776290.1 | 6 - 7 | 1836 | 89 | |
GenBank mRNA | AK089802.1 | - | 6 - 7 | 1837 | 89 | |
BC037033.1 | - | 1 - 2 | 145 | 89 | ||
BC096596.1 | - | 6 - 7 | 1872 | 4239523 | 89 | |
BC116280.1 | - | 6 - 7 | 1644 | 40094408 | 89 | |
BC116281.1 | - | 6 - 7 | 1644 | 40094411 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |