Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc41a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173865.3 | NP_776290.1 | 8 - 9 | 2066 | 61 | |
GenBank mRNA | AK089802.1 | - | 8 - 9 | 2067 | 61 | |
AK154819.1 | - | 8 - 9 | 2041 | 61 | ||
BC037033.1 | - | 3 - 4 | 375 | 61 | ||
BC096596.1 | - | 8 - 9 | 2102 | 4239523 | 61 | |
BC116280.1 | - | 8 - 9 | 1874 | 40094408 | 61 | |
BC116281.1 | - | 8 - 9 | 1874 | 40094411 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |